Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noah-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263093.4 | NP_001250022.1 | 3 - 4 | 1048 | 104 | |
NM_170870.8 | NP_740858.2 | 3 - 4 | 1044 | 104 | ||
NM_171838.7 | NP_740859.3 | 3 - 4 | 1044 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |