Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128939.2 | NP_001122411.1 | 5 - 6 | 607 | 113 | |
NM_001383711.1 | 9 - 10 | 968 | 113 | |||
NM_170918.6 | NP_740912.2 | 8 - 9 | 1379 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |