Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026601.6 | NP_001021772.1 | 7 - 8 | 2558 | 69 | |
NM_001383734.1 | 8 - 9 | 2693 | 69 | |||
NM_170927.5 | NP_740922.1 | 7 - 8 | 2558 | 69 | ||
GenBank mRNA | AF332568.1 | - | 7 - 8 | 2552 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |