Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pll-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171059.6 | NP_741067.1 | 5 - 6 | 564 | 79 | |
NM_171060.5 | NP_741068.1 | 5 - 6 | 592 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |