Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqe-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047389.4 | NP_001040854.1 | 7 - 8 | 774 | 76 | |
NM_001382869.1 | 9 - 10 | 3910 | 76 | |||
NM_065734.8 | NP_498135.2 | 10 - 11 | 3706 | 76 | ||
NM_065735.6 | NP_498136.2 | 7 - 8 | 769 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |