Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306912.2 | NP_001293841.1 | 10 - 11 | 1904 | 136 | |
NM_001306913.3 | NP_001293842.1 | 11 - 12 | 1970 | 136 | ||
NM_001306914.2 | NP_001293843.1 | 10 - 11 | 1868 | 136 | ||
NM_001380288.1 | NP_001368430.1 | 9 - 10 | 1669 | 136 | ||
NM_001383109.1 | 11 - 12 | 1927 | 136 | |||
NM_171367.8 | NP_741430.3 | 10 - 11 | 1801 | 136 | ||
GenBank mRNA | AY455928.1 | - | 10 - 11 | 1863 | 136 | |
AY493413.1 | - | 10 - 11 | 1855 | 136 | ||
AY493414.1 | - | 9 - 10 | 1684 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |