Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380518.2 | NP_001367785.1 | 13 - 14 | 2185 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |