Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rskn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047180.4 | NP_001040645.1 | 6 - 7 | 1482 | 75 | |
NM_001361775.4 | NP_001348666.1 | 6 - 7 | 1484 | 75 | ||
NM_001361776.3 | NP_001348667.1 | 6 - 7 | 1482 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |