Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: del-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059829.6 | NP_492230.2 | 5 - 6 | 518 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |