Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026296.3 | NP_001021467.1 | 8 - 9 | 2485 | 110 | |
NM_001026297.6 | NP_001021468.1 | 8 - 9 | 1360 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |