Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drsh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128988.5 | NP_001122460.2 | 2 - 3 | 680 | 106 | |
NM_060198.5 | NP_492599.1 | 2 - 3 | 678 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |