Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047198.5 | NP_001040663.1 | 9 - 10 | 1686 | 98 | |
NM_001047199.4 | NP_001040664.1 | 2 - 3 | 360 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |