Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060207.6 | NP_492608.2 | 4 - 5 | 442 | 90 | |
GenBank mRNA | AF456248.1 | - | 4 - 5 | 441 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |