Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058458.7 | NP_490859.1 | 9 - 10 | 1845 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |