Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zwl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262221.3 | NP_001249150.1 | 4 - 5 | 794 | 80 | |
NM_001262222.4 | NP_001249151.1 | 4 - 5 | 796 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |