Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373813.1 | NP_001360094.1 | 5 - 6 | 841 | 79 | |
NM_058775.7 | NP_491176.1 | 5 - 6 | 847 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |