Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322672.3 | NP_001309486.1 | 2 - 3 | 234 | 139 | |
NM_001383080.1 | 6 - 7 | 785 | 139 | |||
NM_059174.8 | NP_491575.1 | 6 - 7 | 979 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |