Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059263.10 | NP_491664.1 | 3 - 4 | 335 | 89 | |
GenBank mRNA | U70618.1 | - | 3 - 4 | 277 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |