Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059280.5 | NP_491681.1 | 4 - 5 | 1845 | 68 | |
GenBank mRNA | AF079509.1 | - | 4 - 5 | 1867 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |