Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380377.2 | NP_001367356.1 | 11 - 12 | 3279 | 165 | |
NM_059282.5 | NP_491683.1 | 12 - 13 | 3331 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |