Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059308.8 | NP_491709.1 | 7 - 8 | 1519 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |