Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171399.6 | NP_741465.1 | 9 - 10 | 1201 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |