Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xbx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373079.3 | NP_001359646.1 | 2 | 236 | 73 | |
NM_171511.7 | NP_741597.1 | 3 - 4 | 238 | 73 | ||
NR_069217.1 | 3 - 4 | 241 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |