Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269281.3 | NP_001256210.1 | 6 - 7 | 1179 | 63 | |
NM_171529.6 | NP_741616.2 | 8 - 9 | 1520 | 63 | ||
NM_171530.6 | NP_741617.2 | 7 - 8 | 1263 | 63 | ||
NM_171935.6 | NP_741618.2 | 7 - 8 | 1254 | 63 | ||
NR_132530.1 | 7 - 8 | 1247 | 63 | |||
NR_132531.1 | 7 - 8 | 1254 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |