Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 9 - 10 | 7328 | 102 | |
NM_001269453.3 | NP_001256382.1 | 7 - 8 | 7079 | 102 | ||
NM_001269454.2 | NP_001256383.1 | 9 - 10 | 7570 | 102 | ||
NM_001373124.2 | NP_001360523.1 | 9 | 7637 | 102 | ||
NM_001383415.1 | 9 - 10 | 7576 | 102 | |||
NM_171544.7 | NP_741632.2 | 10 - 11 | 7752 | 102 | ||
GenBank mRNA | AF053496.1 | - | 9 - 10 | 7328 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |