Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029391.4 | NP_001024562.1 | 5 - 6 | 1055 | 76 | |
NM_001381015.1 | NP_001367515.1 | 5 - 6 | 1059 | 76 | ||
GenBank mRNA | AF497826.1 | - | 5 - 6 | 1077 | 76 | |
AF497827.1 | - | 2 - 3 | 797 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |