Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029268.5 | NP_001024439.1 | 10 - 11 | 1289 | 104 | |
NM_171796.5 | NP_741925.2 | 6 - 7 | 1132 | 104 | ||
GenBank mRNA | AJ133838.1 | - | 6 - 7 | 857 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |