Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375204.1 | NP_001361941.1 | 7 - 8 | 897 | 104 | |
NM_171812.7 | NP_741945.3 | 8 - 9 | 1081 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |