Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ajm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330906.2 | NP_001317734.1 | 14 - 15 | 2089 | 71 | |
NM_001330908.3 | NP_001317889.1 | 8 - 9 | 1269 | 71 | ||
NM_001330909.3 | NP_001317890.1 | 8 - 9 | 1044 | 71 | ||
NM_001330910.3 | NP_001317891.1 | 6 - 7 | 912 | 71 | ||
NM_001330911.3 | NP_001317888.1 | 5 - 6 | 798 | 71 | ||
NM_001330912.3 | NP_001317897.1 | 4 - 5 | 482 | 71 | ||
NM_001330913.3 | NP_001317898.1 | 3 - 4 | 237 | 71 | ||
NM_001330914.3 | NP_001317899.1 | 2 - 3 | 165 | 71 | ||
NM_001381045.1 | NP_001368606.1 | 14 - 15 | 2052 | 71 | ||
NM_001381046.2 | NP_001367383.1 | 14 - 15 | 2060 | 71 | ||
NM_001381047.2 | NP_001367382.1 | 14 - 15 | 2052 | 71 | ||
NM_001381048.1 | NP_001367381.1 | 14 - 15 | 2089 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |