Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spe-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136278.3 | NP_001129750.1 | 7 - 8 | 859 | 67 | |
NM_060554.5 | NP_492955.1 | 14 - 15 | 1724 | 67 | ||
GenBank mRNA | AF049327.1 | - | 14 - 15 | 1702 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |