Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060726.1 | NP_493127.1 | 4 - 5 | 1034 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |