Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zoo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265447.3 | NP_001252376.1 | 3 - 4 | 272 | 62 | |
NM_001265448.4 | NP_001252377.1 | 2 - 3 | 175 | 62 | ||
NM_001377679.2 | NP_001364648.1 | 3 - 4 | 274 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |