Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026292.5 | NP_001021463.1 | 11 - 12 | 2718 | 79 | |
NM_061174.6 | NP_493575.2 | 11 - 12 | 2713 | 79 | ||
GenBank mRNA | U33051.1 | - | 11 - 12 | 2714 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |