Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061200.6 | NP_493601.2 | 10 - 11 | 13324 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |