Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269407.2 | NP_001256336.1 | 1 - 2 | 67 | 134 | |
NM_001269408.2 | NP_001256337.1 | 1 - 2 | 120 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |