Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062406.6 | NP_494807.1 | 9 - 10 | 2613 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |