Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313625.4 | NP_001300554.1 | 2 - 3 | 536 | 125 | |
NM_001383826.1 | 3 - 4 | 735 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |