Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383859.1 | 5 - 6 | 1094 | 70 | ||
NR_165322.1 | 5 - 6 | 1040 | 70 | |||
NR_165323.1 | 5 - 6 | 1040 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |