Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062862.5 | NP_495263.2 | 1 - 2 | 289 | 70 |
Gene Symbol: C29F5.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062861.5 | NP_495262.2 | 1 - 2 | 267 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |