Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ani-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062881.4 | NP_495282.1 | 11 - 12 | 3028 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |