Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aff-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063001.6 | NP_495402.3 | 10 - 11 | 1514 | 83 | |
GenBank mRNA | EF205023.1 | - | 10 - 11 | 1514 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |