Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmaa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027131.6 | NP_001022302.1 | 2 - 3 | 1109 | 119 | |
NM_001383882.1 | 2 - 3 | 1120 | 119 |
Gene Symbol: farl-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063066.8 | NP_495467.2 | 10 - 11 | 4442 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |