Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026889.5 | NP_001022060.1 | 8 - 9 | 1196 | 99 | |
NM_001313600.3 | NP_001300529.1 | 7 | 1084 | 99 | ||
NM_001313601.3 | NP_001300530.1 | 7 | 1057 | 99 | ||
NM_001313602.3 | NP_001300531.1 | 6 - 7 | 967 | 99 | ||
NM_001313603.2 | NP_001300532.1 | 5 - 6 | 871 | 99 | ||
NM_001313604.3 | NP_001300533.1 | 7 | 1057 | 99 | ||
NM_001313605.3 | NP_001300534.1 | 6 - 7 | 967 | 99 | ||
NM_001313606.3 | NP_001300535.1 | 7 - 8 | 1084 | 99 | ||
NR_132585.1 | 7 - 8 | 1084 | 99 | |||
GenBank mRNA | AF319614.1 | - | 5 - 6 | 950 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |