Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aka-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026874.4 | NP_001022045.1 | 6 - 7 | 2132 | 89 | |
NM_001383888.1 | 7 - 8 | 2019 | 89 | |||
NM_063164.8 | NP_495565.2 | 6 - 7 | 2144 | 89 | ||
GenBank mRNA | AF045926.1 | - | 6 - 7 | 2143 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |