Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063279.5 | NP_495680.1 | 11 - 12 | 2066 | 78 | |
GenBank mRNA | AF173372.1 | - | 11 - 12 | 2066 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |