Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063312.7 | NP_495713.2 | 12 - 13 | 4897 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |