Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063465.5 | NP_495866.2 | 13 - 14 | 3564 | 96 | |
GenBank mRNA | AB223006.1 | - | 13 - 14 | 3586 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |