Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063539.6 | NP_495940.1 | 3 - 4 | 442 | 71 | |
GenBank mRNA | AB035090.2 | - | 3 - 4 | 440 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |