Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383927.1 | 7 - 8 | 3617 | 66 | ||
NM_001383928.1 | 7 - 8 | 3196 | 66 | |||
GenBank mRNA | AB038503.1 | - | 4 - 5 | 659 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |