Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063857.2 | NP_496258.1 | 2 - 3 | 161 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |